Learn More
Wheaton™ PTFE/Silicone Septa
Brand:  Wheaton™ W240586A
2023.50 DKK valid until 2025-05-31
Use promo code "24127" to get your promotional price.
 View more versions of this product
Product Code. 10033843
  • 2130.00 DKK / Pack of 100
In Stock
to see stock.
Explore more special offers
Specifications
20 mm
Autoclavable
Septa
PTFE, Silicone
100 Pk.
SDS
Documents Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    0
    Hours
    19
    Minutes
    30
    Seconds
    59
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127
    Expires2025-05-31
    Days
    30
    Hours
    4
    Minutes
    29
    Seconds
    0
Product Content Correction
Product Title
Wheaton™ PTFE/Silicone Septa > Outer Diameter: 20mm