Learn More
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
10033843
|
Wheaton™
W240586A |
20 mm |
2130.00 DKK
Pack of 100 |
In Stock
Log in to see stock. |
||||||
12373770
|
Wheaton™
W240581A |
8 mm |
913.00 DKK
Pack of 120 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||||
10151372
|
Wheaton™
W240585 |
18 mm |
1935.00 DKK
Pack of 100 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||||
10210042
|
Wheaton™
W240584A |
15 mm |
1490.00 DKK
Pack of 100 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
11724239
|
Wheaton™
W240583A |
13 mm |
959.00 DKK
Pack of 100 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
Description
Specifications
PTFE, Silicone | |
Autoclavable | |
Septa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Wheaton™ PTFE/Silicone Septa