Learn More
Minebea Intec™ Puro™ Floor Scales
Puro™ floor scales are a sturdy weighing solution for diverse industrial weighing needs.
POA
Specifications
Type | Floor Scale |
---|---|
Display Type | Backlit LCD |
Protection Class | IP43 (Indicator), IP67 (Platform) |
Product Code | Brand | Dimensions (L x W x H) | Includes | Capacity | Readability | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Includes | Capacity | Readability | Price | Quantity & Availability | |||||
17738949
![]() |
Minebea Intec™
EF-4PLLP1500-3d |
1000 x 1000 x 105 mm | Plastic Indicator, Painted Carbon Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display | 1500 kg | 0.5 kg | |||||||
17748949
![]() |
Minebea Intec™
EF-4PLLS1500-3d |
1000 x 1000 x 105 mm | Plastic Indicator, Stainless Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display | 1500 kg | 0.5 kg | |||||||
17758949
![]() |
Minebea Intec™
EF-4PNNP1500-3d |
1250 x 1250 x 105 mm | Plastic Indicator, Painted Carbon Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display | 1500 kg | 0.5 kg | |||||||
17768949
![]() |
Minebea Intec™
EF-4PNNS1500-3d |
1250 x 1250 x 105 mm | Plastic Indicator, Stainless Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display | 1500 kg | 0.5 kg | |||||||
17778949
![]() |
Minebea Intec™
EF-4PRRP1500-3d |
1500 x 1500 x 105 mm | Plastic Indicator, Painted Carbon Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display | 1500 kg | 0.5 kg | |||||||
17798949
![]() |
Minebea Intec™
EF-4PRRS1500-3d |
1500 x 1500 x 105 mm | Plastic Indicator, Painted Carbon Steel Frame, Stainless Steel Load Plate, USB-C Port | 1500 kg | 0.5 kg | |||||||
17788949
![]() |
Minebea Intec™
EF-4PRRP3000-3d |
1500 x 1500 x 105 mm | Plastic Indicator, Painted Carbon Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display | 3000 kg | 1 kg | |||||||
17708959
![]() |
Minebea Intec™
EF-4PRRS3000-3d |
1500 x 1500 x 105 mm | Plastic Indicator, Stainless Steel Frame and Load Plate, USB-C Port, 35 mm Weight Display | 3000 kg | 1 kg | |||||||
Description
Puro™ floor scales unite the most needed applications in one multi-purpose indicator with large display for a comfortable user operating experience.
- Accurate and reliable results in a second
- Combined with the robustness and durability of platforms, floor scales fit comfortably in any environment while offering simple charging and connection technology
- Up to 500 hours continuous use between recharges with 12 hour recharge time
- LCD display with white backlight, 35 mm (1.4 in.) height, 6 digits, 14-segment
- For weighing, counting, check weighing, check counting, and totalization
Specifications
Floor Scale | |
Backlit LCD | |
IP43 (Indicator), IP67 (Platform) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.