Learn More
Cole-Parmer™ PVC Bottle Holders
Mount these bottle holders to a wall, hood or benchtop using attached double-faced tape. Cole-Parmer™ Bottle Holders easily handle most standard wash bottles.
Brand:  Cole-Parmer™ 63526-50
294.06 DKK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11786758
  • 345.00 DKK / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Description
  • Sturdy PVC construction
Specifications
Specifications
Bottle Holder
2
78.7 mm
Wall, Hood, or Benchtop
1 Each
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    31
    Hours
    11
    Minutes
    25
    Seconds
    32
Product Content Correction
Product Title
Cole-Parmer™ PVC Bottle Holders > Holds 2