Learn More
Cole-Parmer™ PVC Bottle Holders
Mount these bottle holders to a wall, hood or benchtop using attached double-faced tape. Cole-Parmer™ Bottle Holders easily handle most standard wash bottles.
345.00 DKK - 518.00 DKK
Specifications
Product Type | Bottle Holder |
---|---|
Mounting | Wall, Hood, or Benchtop |
Quantity | 1 Each |
Diameter (Metric) Cavity | 78.7 mm |
Product Code | Brand | No. of Racks | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Racks | Price | Quantity & Availability | ||||||
11786758
|
Cole-Parmer™
63526-50 |
2 |
345.00 DKK
Each |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
11796758
|
Cole-Parmer™
63526-60 |
3 |
518.00 DKK
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
Description
- Sturdy PVC construction
Specifications
Bottle Holder | |
Wall, Hood, or Benchtop | |
1 Each | |
78.7 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Cole-Parmer™ PVC Bottle Holders