Learn More
Clark Scientific™ PVC Large White Spill Trays
Manufactured from rigid PVC, these large white plastic spill trays are for use outside radiation areas.
208.00 DKK - 408.00 DKK
Specifications
Color | White |
---|---|
Material | PVC |
For Use With (Application) | Outside radiation areas |
Packaging Quantity | 1 Ea. |
Product Code | Brand | Type | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Size | Price | Quantity & Availability | |||||
12988837
|
Clark Scientific™
WPVC large |
Large Spill Tray | 700 x 450 mm |
408.00 DKK
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
12968837
|
Clark Scientific™
WPVC Small |
Small Spill Tray | 475 x 360 mm |
208.00 DKK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||||
Description
- Manufactured from rigid PVC
- Available in yellow for use in radiation areas
- Available in white for use outside radiation areas
- Colorless PVC liners are available
Specifications
White | |
PVC | |
Outside radiation areas | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Clark Scientific™ PVC Large White Spill Trays