Learn More
Pyrex™ Graduated Dropping Funnel with Glass Key
Used to transfer fluids.
Brand: Pyrex 3590/06
This item is not returnable.
View return policy
Description
- Complies with ISO 4800 type 4
- Manufactured from Pyrex borosilicate glass
- Double graduated scale showing approximate volume contained/dispensed
- Graduations in white enamel
- With interchangeable glass key
- Supplied with glass stopper
Specifications
500 mL | |
Yes | |
10mL |
Borosilicate Glass | |
Glass stopper and key | |
Dropping Funnel |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Pyrex™ Graduated Dropping Funnel with Glass Key > Capacity: 500mL
Spot an opportunity for improvement?Share a Content Correction