Learn More
Socorex™ Qualitix™ Sterilization Pouch
Fits any small to mid-size sterilization device and provides easy and effective wrapping of items. Made of robust medical grade paper, they tightly close with self-sealing strip to maintain sterility. Content remains visible through PET/CPP transparent laminated film.
Brand:  Socorex™ 3210.0926
 View more versions of this product
Product Code. 15720679
  • 151.00 DKK / Pack of 200
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Description
  • Robust durable material
  • Quick content identification
  • Large self-sealing strip, easy to close
  • Chevron top seal for easy opening
  • Suitable for steam and EO gas sterilization
  • Two sterilization indicators
  • Five different sizes
  • Conformity to ISO 17665-06, ISO 11135, CE
Specifications
Specifications
Autoclavable
Self-adhesive
90 x 260 mm
ISO 17665-06, ISO 11135, CE
Paper
Videos
  • Socorex Qualitix sterilisation pouches
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Socorex Qualitix sterilisation pouches
    Product Title
    Socorex™ Qualitix™ Sterilization Pouch > 90 x 260 mm