Learn More
Socorex™ Qualitix™ Sterilization Pouch
Fits any small to mid-size sterilization device and provides easy and effective wrapping of items. Made of robust medical grade paper, they tightly close with self-sealing strip to maintain sterility. Content remains visible through PET/CPP transparent laminated film.
89.85 DKK - 836.00 DKK
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | ISO 17665-06, ISO 11135, CE |
Closure Type | Self-adhesive |
Material | Paper |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
15710679
|
Socorex™
3210.0816 |
83 x 160 mm |
89.85 DKK
Pack of 200 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
15720679
|
Socorex™
3210.0926 |
90 x 260 mm |
151.00 DKK
Pack of 200 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15730679
|
Socorex™
3210.1326 |
135 x 260 mm |
229.00 DKK
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||||
15740679
|
Socorex™
3210.1936 |
190 x 360 mm |
442.00 DKK
Pack of 200 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
15750679
|
Socorex™
3210.3040 |
300 x 395 mm |
836.00 DKK
Pack of 200 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
Description
- Robust durable material
- Quick content identification
- Large self-sealing strip, easy to close
- Chevron top seal for easy opening
- Suitable for steam and EO gas sterilization
- Two sterilization indicators
- Five different sizes
- Conformity to ISO 17665-06, ISO 11135, CE
Specifications
Autoclavable | |
ISO 17665-06, ISO 11135, CE | |
Self-adhesive | |
Paper |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Socorex™ Qualitix™ Sterilization Pouch