Learn More
Quickfit™ Allihn Condenser with Ground Cone
Ideal for reflux applications.
Brand:  Quickfit CX7/02/SC
1349.00 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 11338074
  • 1420.00 DKK / Each
Estimated Shipment: 25-07-2025
Explore more special offers
Description
Ideal for reflux applications.
  • Inner bulb sections provide increased cooling surface
  • With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ideal for reflux work
400 mm
Borosilicate Glass
400 mm
19/26
Allihn Condenser
Documents
Product Information
Product Title
Quickfit™ Allihn Condenser with Ground Cone > Cone Size: 19/26; Length: 400mm; Effective Length: 250mm