Learn More
Quickfit™ Allihn Condenser with Ground Cone
Ideal for reflux applications.
1395.00 DKK - 1680.00 DKK
Specifications
Material | Borosilicate Glass |
---|---|
Type | Allihn Condenser |
For Use With (Application) | Ideal for reflux work |
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | ||||||
11338074
|
Quickfit
CX7/02/SC |
19/26 |
1420.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
11348074
|
Quickfit
CX7/03/SC |
24/29 |
1445.00 DKK
Each |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||||
12030073
|
Quickfit
CX7/04/SC |
29/32 |
1445.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
11351845
|
Quickfit
CX7/05/SC |
34/35 |
1395.00 DKK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
11371845
|
Quickfit
CX7/08/SC |
50/42 |
1680.00 DKK
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
11361845
|
Quickfit
CX7/06/SC |
40/38 |
1680.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
Description
Ideal for reflux applications.
- Inner bulb sections provide increased cooling surface
- With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
Borosilicate Glass | |
Allihn Condenser | |
Ideal for reflux work |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Quickfit™ Allihn Condenser with Ground Cone