Learn More
Quickfit™ Allihn Condenser with Ground Cone
Ideal for reflux applications.
Brand: Quickfit CX7/05/SC
1074.03 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
Ideal for reflux applications.
- Inner bulb sections provide increased cooling surface
- With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
Ideal for reflux work | |
335 mm | |
Borosilicate Glass |
335 mm | |
34/35 | |
Allihn Condenser |
Product information
File size: 2.8 MB
Product Content Correction
Product Title
Quickfit™ Allihn Condenser with Ground Cone > Cone Size: 34/35; Length: 335mm; Effective Length: 250mm
Spot an opportunity for improvement?Share a Content Correction