Learn More
Sartorius™ Quintix™ Laboratory Balance with Internal Calibration
Laboratory balance with internal calibration and leveling guided by the indicator.
20345.00 DKK - 31970.00 DKK
Specifications
Certifications/Compliance | Council Directive 2009/23/EC, EN 45501:1992, OIML R76:2006 |
---|---|
Display Type | Color Touchscreen |
Material | Glass |
Model | Electronic Toploading Balances |
Electrical Requirements | 100/240 V, 50/60 Hz |
Description
- Intuitive touchscreen in combination with most precise weighing technology
- Improved workflows with easy-to-use application programs
- Generate GLP-compliant documents in a breeze with 'plug and work' connectivity
- Easy operation of balance without having to read a manual thanks to new user interface and color touchscreen
- Integrated application programs and intuitive user guidance for greater efficiency in your daily lab routine
- Highest accuracy and convenience ensured at all times by the isoCAL fully automatic internal adjustment feature
- Easier record keeping using balance's directly integrated PC functions and connectivity to Sartorius™ lab printer
Specifications
Council Directive 2009/23/EC, EN 45501:1992, OIML R76:2006 | |
Glass | |
100/240 V, 50/60 Hz |
Color Touchscreen | |
Electronic Toploading Balances |