Learn More
Sartorius™ Quintix™ Laboratory Balance with Internal Calibration
Laboratory balance with internal calibration and leveling guided by the indicator.
20345.00 DKK - 31970.00 DKK
Specifications
Certifications/Compliance Council Directive 2009/23/EC, EN 45501:1992, OIML R76:2006
Display Type Color Touchscreen
Material Glass
Model Electronic Toploading Balances
Electrical Requirements 100/240 V, 50/60 Hz
View More Specs
Products 2
Product Code Brand Capacity Readability Stabilization Time Price Quantity & Availability  
Product Code Brand Capacity Readability Stabilization Time Price Quantity & Availability  
17711818
View Documents
Sartorius™
610 g 1 mg 1 s This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17701818
View Documents
Sartorius™
6100 g 1 g 1.5 s This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Intuitive touchscreen in combination with most precise weighing technology
  • Improved workflows with easy-to-use application programs
  • Generate GLP-compliant documents in a breeze with 'plug and work' connectivity
  • Easy operation of balance without having to read a manual thanks to new user interface and color touchscreen
  • Integrated application programs and intuitive user guidance for greater efficiency in your daily lab routine
  • Highest accuracy and convenience ensured at all times by the isoCAL fully automatic internal adjustment feature
  • Easier record keeping using balance's directly integrated PC functions and connectivity to Sartorius™ lab printer
Specifications
Specifications
Council Directive 2009/23/EC, EN 45501:1992, OIML R76:2006
Glass
100/240 V, 50/60 Hz
Color Touchscreen
Electronic Toploading Balances