missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Helix-destabilizing Protein
A DNA sequence encoding the Enterobacteria phage T3 (Bacteriophage T3) Helix-destabilizing protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7414-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_523311.1 | |
Helix-destabilizing Protein | |
Research Use Only | |
2.5 | |
Recombinant Protein | |
E. coli | |
MAGFKKKVYT SGLGTAEPYA YLSKPDYGNE ERGFGNPRGV YKVDLTLSNK DPRCQAMVDE IVKTHEEAYA AAVEEFEANP PQVQRGKKPL TPYEGDMPFF DNGDGTTTFK FKCYASFQDK KTKETKHINL VVVDSKGKKI QEVPIIGGGS KLKVKYSLVP YKWNTAVGAS VKLQLESVML VELATFGGGG EDEWADEVED GGYTASESRQ SRDEQEWQED EHEETPDDDE DF | |
Greater than 90% as determined by SDS-PAGE. |
927413 | |
200 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |