missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Helix-destabilizing Protein
A DNA sequence encoding the Enterobacteria phage T3 (Bacteriophage T3) Helix-destabilizing protein, was expressed in the hosts and tags indicated.
2300.00 DKK - 22245.00 DKK
Specifications
Accession Number | NP_523311.1 |
---|---|
Gene ID (Entrez) | 927413 |
Name | Helix-destabilizing Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098750
|
enQuireBio™
QP7414-ec-10ug |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038760
|
enQuireBio™
QP7414-ec-50ug |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088750
|
enQuireBio™
QP7414-ec-100ug |
100 μg |
6415.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16018760
|
enQuireBio™
QP7414-ec-200ug |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028760
|
enQuireBio™
QP7414-ec-500ug |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008760
|
enQuireBio™
QP7414-ec-1mg |
1 mg |
22245.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_523311.1 | |
Helix-destabilizing Protein | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
927413 | |
Research Use Only | |
2.5 | |
Recombinant Protein | |
E. coli | |
MAGFKKKVYT SGLGTAEPYA YLSKPDYGNE ERGFGNPRGV YKVDLTLSNK DPRCQAMVDE IVKTHEEAYA AAVEEFEANP PQVQRGKKPL TPYEGDMPFF DNGDGTTTFK FKCYASFQDK KTKETKHINL VVVDSKGKKI QEVPIIGGGS KLKVKYSLVP YKWNTAVGAS VKLQLESVML VELATFGGGG EDEWADEVED GGYTASESRQ SRDEQEWQED EHEETPDDDE DF | |
Greater than 90% as determined by SDS-PAGE. |