missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chinese Hamster P4HB / ERBA2L Protein

A DNA sequence encoding the Chinese hamster P4HB / ERBA2L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7857-ec-100ug

 View more versions of this product

Product Code. 16041211

  • 6415.00 DKK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001233622.1
P4HB / ERBA2L Protein
Research Use Only
P4hb
Recombinant Protein
E. coli
DAPEEEDNVL VLKKSNFAEA LAAHNYLLVE FYAPWCGHCK ALAPEYAKAA AKLKAEGSEI RLAKVDATEE SDLAQQYGVR GYPTIKFFKN GDTASPKEYT AGREADDIVN WLKKRTGPAA TTLSDTAAAE TLIDSSEVAV IGFFKDVESD SAKQFLLAAE AVDDIPFGIT SNSGVFSKYQ LDKDGVVLFK KFDEGRNNFE GEVTKEKLLD FIKHNQLPLV IEFTEQTAPK IFGGEIKTHI LLFLPKSVSD YDGKLGNFKK AAEGFKGKIL FIFIDSDHTD NQRILEFFGL KKEECPAVRL ITLEEEMTKY KPESDELTAE KITEFCHRFL EGKIKPHLMS QELPEDWDKQ PVKVLVGKNF EEVAFDEKKN VFVEFYAPWC GHCKQLAPIW DKLGETYKDH ENIIIAKMDS TANEVEAVKV HSFPTLKFFP ATADRTVIDY NGERTLDGFK KFLESGGQDG AGDDDDVDLE EALEPDMEED DDQKAVKDEL
Greater than 90% as determined by SDS-PAGE.
100689433
100 μg
Not determined.
Full Length
Chinese Hamster
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers