missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chinese Hamster P4HB / ERBA2L Protein
A DNA sequence encoding the Chinese hamster P4HB / ERBA2L, was expressed in the hosts and tags indicated.
Specifications
Accession Number | NP_001233622.1 |
---|---|
Gene ID (Entrez) | 100689433 |
Name | P4HB / ERBA2L Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051211
|
enQuireBio™
QP7857-ec-10ug |
10 μg |
N/A
|
N/A | |||||
16091211
|
enQuireBio™
QP7857-ec-50ug |
50 μg |
N/A
|
N/A | |||||
16041211
|
enQuireBio™
QP7857-ec-100ug |
100 μg |
N/A
|
N/A | |||||
16071211
|
enQuireBio™
QP7857-ec-200ug |
200 μg |
N/A
|
N/A | |||||
16081211
|
enQuireBio™
QP7857-ec-500ug |
500 μg |
N/A
|
N/A | |||||
16061211
|
enQuireBio™
QP7857-ec-1mg |
1 mg |
N/A
|
N/A | |||||
Specifications
NP_001233622.1 | |
P4HB / ERBA2L Protein | |
Not determined. | |
Full Length | |
Chinese Hamster | |
Tris-based buffer, 50% glycerol |
100689433 | |
Research Use Only | |
P4hb | |
Recombinant Protein | |
DAPEEEDNVL VLKKSNFAEA LAAHNYLLVE FYAPWCGHCK ALAPEYAKAA AKLKAEGSEI RLAKVDATEE SDLAQQYGVR GYPTIKFFKN GDTASPKEYT AGREADDIVN WLKKRTGPAA TTLSDTAAAE TLIDSSEVAV IGFFKDVESD SAKQFLLAAE AVDDIPFGIT SNSGVFSKYQ LDKDGVVLFK KFDEGRNNFE GEVTKEKLLD FIKHNQLPLV IEFTEQTAPK IFGGEIKTHI LLFLPKSVSD YDGKLGNFKK AAEGFKGKIL FIFIDSDHTD NQRILEFFGL KKEECPAVRL ITLEEEMTKY KPESDELTAE KITEFCHRFL EGKIKPHLMS QELPEDWDKQ PVKVLVGKNF EEVAFDEKKN VFVEFYAPWC GHCKQLAPIW DKLGETYKDH ENIIIAKMDS TANEVEAVKV HSFPTLKFFP ATADRTVIDY NGERTLDGFK KFLESGGQDG AGDDDDVDLE EALEPDMEED DDQKAVKDEL |