missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASGPR1 / ASGR1 Protein
A DNA sequence encoding the Homo sapiens (Human) ASGPR1 / ASGR1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8899-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_001184145.1 | |
ASGPR1 / ASGR1 Protein | |
Research Use Only | |
ASGR1 | |
Recombinant Protein | |
E. coli | |
EELRGLRETF SNFTASTEAQ VKGLSTQGGN VGRKMKSLES QLEKQQKDLS EDHSSLLLHV KQFVSDLRSL SCQMAALQGN GSERTCCPVN WVEHERSCYW FSRSGKAWAD ADNYCRLEDA HLVVVTSWEE QKFVQHHIGP VNTWMGLHDQ NGPWKWVDGT DYETGFKNWR PEQPDDWYGH GLGGGEDCAH FTDDGRWNDD VCQRPYRWVC ETELDKASQE PPLL | |
Greater than 90% as determined by SDS-PAGE. |
432 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |