missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASGPR1 / ASGR1 Protein
A DNA sequence encoding the Homo sapiens (Human) ASGPR1 / ASGR1, was expressed in the hosts and tags indicated.
1385.00 DKK - 13770.00 DKK
Specifications
Accession Number | NP_001184145.1 |
---|---|
Gene ID (Entrez) | 432 |
Name | ASGPR1 / ASGR1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087071
|
enQuireBio™
QP8899-ec-10ug |
10 μg |
1385.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16027081
|
enQuireBio™
QP8899-ec-50ug |
50 μg | N/A | ||||||
16077071
|
enQuireBio™
QP8899-ec-100ug |
100 μg | N/A | ||||||
16007081
|
enQuireBio™
QP8899-ec-200ug |
200 μg |
5675.00 DKK
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16017081
|
enQuireBio™
QP8899-ec-500ug |
500 μg | N/A | ||||||
16097071
|
enQuireBio™
QP8899-ec-1mg |
1 mg | N/A | ||||||
Specifications
NP_001184145.1 | |
ASGPR1 / ASGR1 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
432 | |
Research Use Only | |
ASGR1 | |
Recombinant Protein | |
E. coli | |
EELRGLRETF SNFTASTEAQ VKGLSTQGGN VGRKMKSLES QLEKQQKDLS EDHSSLLLHV KQFVSDLRSL SCQMAALQGN GSERTCCPVN WVEHERSCYW FSRSGKAWAD ADNYCRLEDA HLVVVTSWEE QKFVQHHIGP VNTWMGLHDQ NGPWKWVDGT DYETGFKNWR PEQPDDWYGH GLGGGEDCAH FTDDGRWNDD VCQRPYRWVC ETELDKASQE PPLL | |
Greater than 90% as determined by SDS-PAGE. |