missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fibroleukin Protein

A DNA sequence encoding the Homo sapiens (Human) Fibroleukin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7571-ye-500ug

 View more versions of this product

Product Code. 16029480

  • 12590.00 DKK / 500µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_006673.1
Fibroleukin Protein
Research Use Only
FGL2
Recombinant Protein
Yeast
NNETEEIKDE RAKDVCPVRL ESRGKCEEAG ECPYQVSLPP LTIQLPKQFS RIEEVFKEVQ NLKEIVNSLK KSCQDCKLQA DDNGDPGRNG LLLPSTGAPG EVGDNRVREL ESEVNKLSSE LKNAKEEINV LHGRLEKLNL VNMNNIENYV DSKVANLTFV VNSLDGKCSK CPSQEQIQSR PVQHLIYKDC SDYYAIGKRS SETYRVTPDP KNSSFEVYCD METMGGGWTV LQARLDGSTN FTRTWQDYKA GFGNLRREFW LGNDKIHLLT KSKEMILRID LEDFNGVELY ALYDQFYVAN EFLKYRLHVG NYNGTAGDAL RFNKHYNHDL KFFTTPDKDN DRYPSGNCGL YYSSGWWFDA CLSANLNGKY YHQKYRGVRN GIFWGTWPGV SEAHPGGYKS SFKEAKMMIR PKHFKP
Greater than 90% as determined by SDS-PAGE.
10875
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers