missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fibroleukin Protein
A DNA sequence encoding the Homo sapiens (Human) Fibroleukin, was expressed in the hosts and tags indicated.
1500.00 DKK - 15995.00 DKK
Specifications
Accession Number | NP_006673.1 |
---|---|
Gene ID (Entrez) | 10875 |
Name | Fibroleukin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099470
|
enQuireBio™
QP7571-ye-10ug |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039480
|
enQuireBio™
QP7571-ye-50ug |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089470
|
enQuireBio™
QP7571-ye-100ug |
100 μg |
4190.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16019480
|
enQuireBio™
QP7571-ye-200ug |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029480
|
enQuireBio™
QP7571-ye-500ug |
500 μg |
11360.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16009480
|
enQuireBio™
QP7571-ye-1mg |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006673.1 | |
Fibroleukin Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
10875 | |
Research Use Only | |
FGL2 | |
Recombinant Protein | |
Yeast | |
NNETEEIKDE RAKDVCPVRL ESRGKCEEAG ECPYQVSLPP LTIQLPKQFS RIEEVFKEVQ NLKEIVNSLK KSCQDCKLQA DDNGDPGRNG LLLPSTGAPG EVGDNRVREL ESEVNKLSSE LKNAKEEINV LHGRLEKLNL VNMNNIENYV DSKVANLTFV VNSLDGKCSK CPSQEQIQSR PVQHLIYKDC SDYYAIGKRS SETYRVTPDP KNSSFEVYCD METMGGGWTV LQARLDGSTN FTRTWQDYKA GFGNLRREFW LGNDKIHLLT KSKEMILRID LEDFNGVELY ALYDQFYVAN EFLKYRLHVG NYNGTAGDAL RFNKHYNHDL KFFTTPDKDN DRYPSGNCGL YYSSGWWFDA CLSANLNGKY YHQKYRGVRN GIFWGTWPGV SEAHPGGYKS SFKEAKMMIR PKHFKP | |
Greater than 90% as determined by SDS-PAGE. |