missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GSTZ1 Protein
A DNA sequence encoding the Homo sapiens (Human) GSTZ1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6128-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001299589.1 | |
GSTZ1 Protein | |
Research Use Only | |
GSTZ1 | |
Recombinant Protein | |
E. coli | |
MQAGKPILYS YFRSSCSWRV RIALALKGID YETVPINLIK DGGQQFSKDF QALNPMKQVP TLKIDGITIH QSLAIIEYLE EMRPTPRLLP QDPKKRASVR MISDLIAGGI QPLQNLSVLK QVGEEMQLTW AQNAITCGFN ALEQILQSTA GIYCVGDEVT MADLCLVPQV ANAERFKVDL TPYPTISSIN KRLLVLEAFQ VSHPCRQPDT PTELRA | |
Greater than 80% as determined by SDS-PAGE. |
2954 | |
200 μg | |
Not determined. | |
Full Length of BC001453 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |