missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GSTZ1 Protein

A DNA sequence encoding the Homo sapiens (Human) GSTZ1, was expressed in the hosts and tags indicated.

1185.00 DKK - 15565.00 DKK

Specifications

Accession Number NP_001299589.1
Gene ID (Entrez) 2954
Name GSTZ1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16081010
View Documents Promotion Details
enQuireBio™
QP6128-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16021020
View Documents Promotion Details
enQuireBio™
QP6128-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16071010
View Documents Promotion Details
enQuireBio™
QP6128-ec-100ug
100 μg
3450.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16001020
View Documents Promotion Details
enQuireBio™
QP6128-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16011020
View Documents Promotion Details
enQuireBio™
QP6128-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16091010
View Documents Promotion Details
enQuireBio™
QP6128-ec-1mg
1 mg
15565.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001299589.1
GSTZ1 Protein
Not determined.
Full Length of BC001453
Human
GST
Tris-based buffer, 50% glycerol
2954
Research Use Only
GSTZ1
Recombinant Protein
E. coli
MQAGKPILYS YFRSSCSWRV RIALALKGID YETVPINLIK DGGQQFSKDF QALNPMKQVP TLKIDGITIH QSLAIIEYLE EMRPTPRLLP QDPKKRASVR MISDLIAGGI QPLQNLSVLK QVGEEMQLTW AQNAITCGFN ALEQILQSTA GIYCVGDEVT MADLCLVPQV ANAERFKVDL TPYPTISSIN KRLLVLEAFQ VSHPCRQPDT PTELRA
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications