missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Negative elongation factor B Protein

A DNA sequence encoding the Homo sapiens (Human) Negative elongation factor B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8455-ec-1mg

 View more versions of this product

Product Code. 16004721

  • 15565.00 DKK / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_056271.3
Negative elongation factor B Protein
Research Use Only
COBRA1
Recombinant Protein
E. coli
LGVANGEDLK ETLTNCTEPL KAIEQFQTEN GVLLPSLQSA LPFLDLHGTP RLEFHQSVFD ELRDKLLERV SAIASEGKAE ERYKKLEDLL EKSFSLVKMP SLQPVVMCVM KHLPKVPEKK LKLVMADKEL YRACAVEVKR QIWQDNQALF GDEVSPLLKQ YILEKESALF STELSVLHNF FSPSPKTRRQ GE
Greater than 90% as determined by SDS-PAGE.
25920
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers