missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Negative elongation factor B Protein
A DNA sequence encoding the Homo sapiens (Human) Negative elongation factor B, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_056271.3 |
---|---|
Gene ID (Entrez) | 25920 |
Name | Negative elongation factor B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094711
|
enQuireBio™
QP8455-ec-10ug |
10 μg | N/A | ||||||
16034721
|
enQuireBio™
QP8455-ec-50ug |
50 μg | N/A | ||||||
16084711
|
enQuireBio™
QP8455-ec-100ug |
100 μg |
3450.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16014721
|
enQuireBio™
QP8455-ec-200ug |
200 μg | N/A | ||||||
16024721
|
enQuireBio™
QP8455-ec-500ug |
500 μg | N/A | ||||||
16004721
|
enQuireBio™
QP8455-ec-1mg |
1 mg |
15565.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_056271.3 | |
Negative elongation factor B Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
25920 | |
Research Use Only | |
COBRA1 | |
Recombinant Protein | |
E. coli | |
LGVANGEDLK ETLTNCTEPL KAIEQFQTEN GVLLPSLQSA LPFLDLHGTP RLEFHQSVFD ELRDKLLERV SAIASEGKAE ERYKKLEDLL EKSFSLVKMP SLQPVVMCVM KHLPKVPEKK LKLVMADKEL YRACAVEVKR QIWQDNQALF GDEVSPLLKQ YILEKESALF STELSVLHNF FSPSPKTRRQ GE | |
Greater than 90% as determined by SDS-PAGE. |