missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDZ and LIM domain protein 1

A DNA sequence encoding the Homo sapiens (Human) PDZ and LIM domain protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7666-ec-200ug

 View more versions of this product

Product Code. 16080071

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001107579.1
PDZ and LIM domain protein 1
Research Use Only
PDLIM3
Recombinant Protein
E. coli
MPQTVILPGP APWGFRLSGG IDFNQPLVIT RITPGSKAAA ANLCPGDVIL AIDGFGTESM THADAQDRIK AAAHQLCLKI DRGETHLWSP QVSEDGKAHP FKINLESEPQ EFKPIGTAHN RRAQPFVAAA NIDDKRQVVS ASYNSPIGLY STSNIQDALH GQLRGLIPSS PQKTGTTLNT SIIFGPNLS
Greater than 90% as determined by SDS-PAGE.
27295
200 μg
Not determined.
Full Length of Isoform 3
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers