missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDZ and LIM domain protein 1
A DNA sequence encoding the Homo sapiens (Human) PDZ and LIM domain protein 3, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_001107579.1 |
---|---|
Gene ID (Entrez) | 27295 |
Name | PDZ and LIM domain protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16060071
|
enQuireBio™
QP7666-ec-10ug |
10 μg | N/A | ||||||
16000081
|
enQuireBio™
QP7666-ec-50ug |
50 μg | N/A | ||||||
16050071
|
enQuireBio™
QP7666-ec-100ug |
100 μg |
3450.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16080071
|
enQuireBio™
QP7666-ec-200ug |
200 μg | N/A | ||||||
16090071
|
enQuireBio™
QP7666-ec-500ug |
500 μg | N/A | ||||||
16070071
|
enQuireBio™
QP7666-ec-1mg |
1 mg |
15565.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001107579.1 | |
PDZ and LIM domain protein 1 | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
27295 | |
Research Use Only | |
PDLIM3 | |
Recombinant Protein | |
E. coli | |
MPQTVILPGP APWGFRLSGG IDFNQPLVIT RITPGSKAAA ANLCPGDVIL AIDGFGTESM THADAQDRIK AAAHQLCLKI DRGETHLWSP QVSEDGKAHP FKINLESEPQ EFKPIGTAHN RRAQPFVAAA NIDDKRQVVS ASYNSPIGLY STSNIQDALH GQLRGLIPSS PQKTGTTLNT SIIFGPNLS | |
Greater than 90% as determined by SDS-PAGE. |