missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Retinol-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) Retinol-binding protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6588-ec-100ug

 View more versions of this product

Product Code. 16044360

  • 3825.00 DKK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_004155.2
Retinol-binding protein 1
Research Use Only
RBP2
Recombinant Protein
E. coli
TRDQNGTWEM ESNENFEGYM KALDIDFATR KIAVRLTQTK VIDQDGDNFK TKTTSTFRNY DVDFTVGVEF DEYTKSLDNR HVKALVTWEG DVLVCVQKGE KENRGWKQWI EGDKLYLELT CGDQVCRQVF KKK
Greater than 80% as determined by SDS-PAGE.
5948
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers