missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Retinol-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Retinol-binding protein 2, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_004155.2 |
---|---|
Gene ID (Entrez) | 5948 |
Name | Retinol-binding protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16054360
|
enQuireBio™
QP6588-ec-10ug |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094360
|
enQuireBio™
QP6588-ec-50ug |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044360
|
enQuireBio™
QP6588-ec-100ug |
100 μg |
3450.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16074360
|
enQuireBio™
QP6588-ec-200ug |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084360
|
enQuireBio™
QP6588-ec-500ug |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064360
|
enQuireBio™
QP6588-ec-1mg |
1 mg |
15565.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_004155.2 | |
Retinol-binding protein 1 | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5948 | |
Research Use Only | |
RBP2 | |
Recombinant Protein | |
E. coli | |
TRDQNGTWEM ESNENFEGYM KALDIDFATR KIAVRLTQTK VIDQDGDNFK TKTTSTFRNY DVDFTVGVEF DEYTKSLDNR HVKALVTWEG DVLVCVQKGE KENRGWKQWI EGDKLYLELT CGDQVCRQVF KKK | |
Greater than 80% as determined by SDS-PAGE. |