missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Retinol-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) Retinol-binding protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6588-ec-200ug

 View more versions of this product

Product Code. 16074360

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_004155.2
Retinol-binding protein 1
Research Use Only
RBP2
Recombinant Protein
E. coli
TRDQNGTWEM ESNENFEGYM KALDIDFATR KIAVRLTQTK VIDQDGDNFK TKTTSTFRNY DVDFTVGVEF DEYTKSLDNR HVKALVTWEG DVLVCVQKGE KENRGWKQWI EGDKLYLELT CGDQVCRQVF KKK
Greater than 80% as determined by SDS-PAGE.
5948
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales