missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Serine / threonine-protein kinase NIM1 Protein

A DNA sequence encoding the Homo sapiens (Human) Serine/threonine-protein kinase NIM1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7811-ec-10ug

 View more versions of this product

Product Code. 16080941

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_699192.1
Serine / threonine-protein kinase NIM1 Protein
Research Use Only
NIM1
Recombinant Protein
E. coli
MTAVYMNGGG LVNPHYARWD RRDSVESGCQ TESSKEGEEG QPRQLTPFEK LTQDMSQDEK VVREITLGKR IGFYRIRGEI GSGNFSQVKL GIHSLTKEKV AIKILDKTKL DQKTQRLLSR EISSMEKLHH PNIIRLYEVV ETLSKLHLVM EYAGGGELFG KISTEGKLSE PESKLIFSQI VSAVKHMHEN QIIHRDLKAE NVFYTSNTCV KVGDFGFSTV SKKGEMLNTF CGSPPYAAPE LFRDEHYIGI YVDIWALGVL LYFMVTGTMP FRAETVAKLK KSILEGTYSV PPHVSEPCHR LIRGVLQQIP TERYGIDCIM NDEWMQGVPY PTPLEPFQLD PKHLSETSTL KEEENEVKST LEHLGITEEH IRNNQGRDAR SSITGVYRII LHRVQRKKAL ESVPVMMLPD PKERDLKKGS RVYRGIRHTS KFCSIL
Greater than 90% as determined by SDS-PAGE.
167359
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers