missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine / threonine-protein kinase NIM1 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/threonine-protein kinase NIM1, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_699192.1 |
---|---|
Gene ID (Entrez) | 167359 |
Name | Serine / threonine-protein kinase NIM1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16080941
|
enQuireBio™
QP7811-ec-10ug |
10 μg | N/A | ||||||
16020951
|
enQuireBio™
QP7811-ec-50ug |
50 μg | N/A | ||||||
16070941
|
enQuireBio™
QP7811-ec-100ug |
100 μg |
3450.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16000951
|
enQuireBio™
QP7811-ec-200ug |
200 μg | N/A | ||||||
16010951
|
enQuireBio™
QP7811-ec-500ug |
500 μg | N/A | ||||||
16090941
|
enQuireBio™
QP7811-ec-1mg |
1 mg |
15565.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_699192.1 | |
Serine / threonine-protein kinase NIM1 Protein | |
Not determined. | |
Full Length | |
Human | |
Tris-based buffer, 50% glycerol |
167359 | |
Research Use Only | |
NIM1 | |
Recombinant Protein | |
MTAVYMNGGG LVNPHYARWD RRDSVESGCQ TESSKEGEEG QPRQLTPFEK LTQDMSQDEK VVREITLGKR IGFYRIRGEI GSGNFSQVKL GIHSLTKEKV AIKILDKTKL DQKTQRLLSR EISSMEKLHH PNIIRLYEVV ETLSKLHLVM EYAGGGELFG KISTEGKLSE PESKLIFSQI VSAVKHMHEN QIIHRDLKAE NVFYTSNTCV KVGDFGFSTV SKKGEMLNTF CGSPPYAAPE LFRDEHYIGI YVDIWALGVL LYFMVTGTMP FRAETVAKLK KSILEGTYSV PPHVSEPCHR LIRGVLQQIP TERYGIDCIM NDEWMQGVPY PTPLEPFQLD PKHLSETSTL KEEENEVKST LEHLGITEEH IRNNQGRDAR SSITGVYRII LHRVQRKKAL ESVPVMMLPD PKERDLKKGS RVYRGIRHTS KFCSIL |