missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Stabilin-2 (aa 2198-2295) Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 3645-S2-025
3383.06 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Stabilin-2 (aa 2198-2295) Protein is derived from E. coli. The Recombinant Human Stabilin-2 (aa 2198-2295) Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
12.6 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
55576 | |
Stabilin-2 | |
E. coli-derived human Stabilin-2 protein, (N-terminus) MNHKVHM Human Stabilin (Gly2198-Val2295) Accession &Num; Q8WWQ8 6-His tag (C-terminus) | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |