missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Stabilin-2 (aa 2198-2295) Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
4295.00 DKK
Specifications
Gene ID (Entrez) | 55576 |
---|---|
Molecular Weight (g/mol) | 12.6 kDa |
Name | Stabilin-2 |
Quantity | 25 μg |
Source | E. coli-derived human Stabilin-2 protein, (N-terminus) MNHKVHM Human Stabilin (Gly2198-Val2295) Accession &Num; Q8WWQ8 6-His tag (C-terminus) |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18167810
|
R&D Systems™
3645-S2-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
4295.00 DKK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18186380
|
R&D Systems™
3645-S2-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
4295.00 DKK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human Stabilin-2 (aa 2198-2295) Protein is derived from E. coli. The Recombinant Human Stabilin-2 (aa 2198-2295) Protein has been validated for the following applications: Binding Activity.Specifications
55576 | |
Stabilin-2 | |
E. coli-derived human Stabilin-2 protein, (N-terminus) MNHKVHM Human Stabilin (Gly2198-Val2295) Accession &Num; Q8WWQ8 6-His tag (C-terminus) | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
12.6 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |