Learn More
enQuireBio™ Recombinant Human UDP-glucose 4-epimerase Protein
A DNA sequence encoding the Homo sapiens (Human) UDP-glucose 4-epimerase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7531-ec-1mg
Specifications
NP_000394.2 | |
UDP-glucose 4-epimerase Protein | |
Research Use Only | |
GALE | |
Recombinant Protein | |
E. coli | |
MAEKVLVTGG AGYIGSHTVL ELLEAGYLPV VIDNFHNAFR GGGSLPESLR RVQELTGRSV EFEEMDILDQ GALQRLFKKY SFMAVIHFAG LKAVGESVQK PLDYYRVNLT GTIQLLEIMK AHGVKNLVFS SSATVYGNPQ YLPLDEAHPT GGCTNPYGKS KFFIEEMIRD LCQADKTWNA VLLRYFNPTG AHASGCIGED PQGIPNNLMP YVSQVAIGRR EALNVFGNDY DTEDGTGVRD YIHVVDLAKG HIAALRKLKE QCGCRIYNLG TGTGYSVLQM VQAMEKASGK KIPYKVVARR EGDVAACYAN PSLAQEELGW TAALGLDRMC EDLWRWQKQN PSGFGTQA | |
Greater than 90% as determined by SDS-PAGE. |
2582 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.