missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H2N2 (Mallard / New York / 6750 / 1978) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (strain A/Mallard/New York/6750/1978 H2N2) Influenza A H2N2 (strain A/Mallard/New York/6750/1978) Hemagglutinin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7702-ec-10ug
This item is not returnable.
View return policy
Specifications
Influenza A H2N2 (Mallard / New York / 6750 / 1978) Hemagglutinin Protein | |
Research Use Only | |
HA | |
Recombinant Protein | |
E. coli | |
DQICIGYHAN NSTEKVDTIL ERNVTVTHAK DILEKTHNGK LCRLSGIPPL ELGDCSIAGW LLGNPECDRL LSVPEWSYIV EKENPANGLC YPGNFNDYEE LKHLLTRVTH FEKIKILPRD QWTQHTTTGG SRACAVSGNP SFFRNMVWLT KKGSNYPVAK GSYNNTSGEQ MLVIWGIHHP NDDTEQRTLY QNVGTYVSVG TSTLNKRSIP EIATRPKVNG QGGRMEFSWT LLETWDVINF ESTGNLIAPE YGFKISKRGS SGIMKTEKTL ENCETKCQTP LGAINTTLPF HNIHPLTIGE CPKYVKSDRL VLATGLRNVP QIES | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |