missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H2N2 (Mallard / New York / 6750 / 1978) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (strain A/Mallard/New York/6750/1978 H2N2) Influenza A H2N2 (strain A/Mallard/New York/6750/1978) Hemagglutinin, was expressed in the hosts and tags indicated.
2300.00 DKK - 22245.00 DKK
Specifications
Name | Influenza A H2N2 (Mallard / New York / 6750 / 1978) Hemagglutinin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | HA |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16090281
|
enQuireBio™
QP7702-ec-10ug |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030291
|
enQuireBio™
QP7702-ec-50ug |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080281
|
enQuireBio™
QP7702-ec-100ug |
100 μg |
6415.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16010291
|
enQuireBio™
QP7702-ec-200ug |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020291
|
enQuireBio™
QP7702-ec-500ug |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000291
|
enQuireBio™
QP7702-ec-1mg |
1 mg |
22245.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Influenza A H2N2 (Mallard / New York / 6750 / 1978) Hemagglutinin Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Influenza | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
HA | |
Recombinant Protein | |
DQICIGYHAN NSTEKVDTIL ERNVTVTHAK DILEKTHNGK LCRLSGIPPL ELGDCSIAGW LLGNPECDRL LSVPEWSYIV EKENPANGLC YPGNFNDYEE LKHLLTRVTH FEKIKILPRD QWTQHTTTGG SRACAVSGNP SFFRNMVWLT KKGSNYPVAK GSYNNTSGEQ MLVIWGIHHP NDDTEQRTLY QNVGTYVSVG TSTLNKRSIP EIATRPKVNG QGGRMEFSWT LLETWDVINF ESTGNLIAPE YGFKISKRGS SGIMKTEKTL ENCETKCQTP LGAINTTLPF HNIHPLTIGE CPKYVKSDRL VLATGLRNVP QIES |