missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) 2-deoxyglucose-6-phosphate phosphatase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9588-ye-1mg
This item is not returnable.
View return policy
Specifications
NP_011910.1 | |
S. cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 Protein | |
Research Use Only | |
DOG1 | |
Recombinant Protein | |
Yeast | |
MAEFSADLCL FDLDGTIVST TVAAEKAWTK LCYEYGVDPS ELFKHSHGAR TQEVLRRFFP KLDDTDNKGV LALEKDIAHS YLDTVSLIPG AENLLLSLDV DTETQKKLPE RKWAIVTSGS PYLAFSWFET ILKNVGKPKV FITGFDVKNG KPDPEGYSRA RDLLRQDLQL TGKQDLKYVV FEDAPVGIKA GKAMGAITVG ITSSYDKSVL FDAGADYVVC DLTQVSVVKN NENGIVIQVN NPLTRA | |
Greater than 85.6% as determined by SDS-PAGE. |
856440 | |
1 mg | |
Not determined. | |
Full. Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |