missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) 2-deoxyglucose-6-phosphate phosphatase 1, was expressed in the hosts and tags indicated.

2615.00 DKK - 23570.00 DKK

Specifications

Accession Number NP_011910.1
Gene ID (Entrez) 856440
Name S. cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16038501
View Documents Promotion Details
enQuireBio™
QP9588-ye-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16078501
View Documents Promotion Details
enQuireBio™
QP9588-ye-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16028501
View Documents Promotion Details
enQuireBio™
QP9588-ye-100ug
100 μg
7160.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16058501
View Documents Promotion Details
enQuireBio™
QP9588-ye-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16068501
View Documents Promotion Details
enQuireBio™
QP9588-ye-500ug
500 μg
17300.00 DKK
500µg
Please to purchase this item. Need a web account? Register with us today!
16048501
View Documents Promotion Details
enQuireBio™
QP9588-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_011910.1
S. cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 Protein
Not determined.
Full. Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
856440
Research Use Only
DOG1
Recombinant Protein
Yeast
MAEFSADLCL FDLDGTIVST TVAAEKAWTK LCYEYGVDPS ELFKHSHGAR TQEVLRRFFP KLDDTDNKGV LALEKDIAHS YLDTVSLIPG AENLLLSLDV DTETQKKLPE RKWAIVTSGS PYLAFSWFET ILKNVGKPKV FITGFDVKNG KPDPEGYSRA RDLLRQDLQL TGKQDLKYVV FEDAPVGIKA GKAMGAITVG ITSSYDKSVL FDAGADYVVC DLTQVSVVKN NENGIVIQVN NPLTRA
Greater than 85.6% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications