missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Pyruvate carboxylase 1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Pyruvate carboxylase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7045-ye-50ug
This item is not returnable.
View return policy
Specifications
NP_011453.1 | |
S. cerevisiae Pyruvate carboxylase 1 Protein | |
Research Use Only | |
PYC1 | |
Recombinant Protein | |
Yeast | |
EKNKILVANR GEIPIRIFRT AHELSMQTVA IYSHEDRLST HKQKADEAYV IGEVGQYTPV GAYLAIDEII SIAQKHQVDF IHPGYGFLSE NSEFADKVVK AGITWIGPPA EVIDSVGDKV SARNLAAKAN VPTVPGTPGP IETVEEALDF VNEYGYPVII KAAFGGGGRG MRVVREGDDV ADAFQRATSE ARTAFGNGTC FVERFLDKPK HIEVQLLADN HGNVVHLFER DCSVQRRHQK VVEVAPAKTL PREVRDAILT DAVKLAKECG YRNAGTAEFL VDNQNRHYFI EINPRIQVEH TITEEITGID IVAAQIQIAA GASLPQLGLF QDKITTRGFA IQCRITTEDP AKNFQPDTGR IEVYRSAGGN GVRLDGGNAY AGTIISPHYD SMLVKCSCSG STYEIVRRKM IRALIEFRIR GVKTNIPFLL TLLTNPVFIE GTYWTTFIDD TPQ | |
Greater than 90% as determined by SDS-PAGE. |
852818 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |