missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Pyruvate carboxylase 1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Pyruvate carboxylase 1, was expressed in the hosts and tags indicated.
2615.00 DKK - 23570.00 DKK
Specifications
Accession Number | NP_011453.1 |
---|---|
Gene ID (Entrez) | 852818 |
Name | S. cerevisiae Pyruvate carboxylase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037160
|
enQuireBio™
QP7045-ye-10ug |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077160
|
enQuireBio™
QP7045-ye-50ug |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027160
|
enQuireBio™
QP7045-ye-100ug |
100 μg |
7160.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16057160
|
enQuireBio™
QP7045-ye-200ug |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067160
|
enQuireBio™
QP7045-ye-500ug |
500 μg |
17300.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16047160
|
enQuireBio™
QP7045-ye-1mg |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_011453.1 | |
S. cerevisiae Pyruvate carboxylase 1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |
852818 | |
Research Use Only | |
PYC1 | |
Recombinant Protein | |
Yeast | |
EKNKILVANR GEIPIRIFRT AHELSMQTVA IYSHEDRLST HKQKADEAYV IGEVGQYTPV GAYLAIDEII SIAQKHQVDF IHPGYGFLSE NSEFADKVVK AGITWIGPPA EVIDSVGDKV SARNLAAKAN VPTVPGTPGP IETVEEALDF VNEYGYPVII KAAFGGGGRG MRVVREGDDV ADAFQRATSE ARTAFGNGTC FVERFLDKPK HIEVQLLADN HGNVVHLFER DCSVQRRHQK VVEVAPAKTL PREVRDAILT DAVKLAKECG YRNAGTAEFL VDNQNRHYFI EINPRIQVEH TITEEITGID IVAAQIQIAA GASLPQLGLF QDKITTRGFA IQCRITTEDP AKNFQPDTGR IEVYRSAGGN GVRLDGGNAY AGTIISPHYD SMLVKCSCSG STYEIVRRKM IRALIEFRIR GVKTNIPFLL TLLTNPVFIE GTYWTTFIDD TPQ | |
Greater than 90% as determined by SDS-PAGE. |