Learn More
Gilac™ Reinforced Stacking and Nesting Container
Reinforced Stacking and Nesting Container is designed for intensive use and extreme handling conditions; it's stackable and nestable by a quarter turn.
Brand: Gilac™ G169321
This item is not returnable.
View return policy
Description
- Reinforced containers for intensive use and extreme handling conditions.
- Stackable and nestable by a quarter turn.
- Highly durable.
- Suitable for a number of different uses in the food processing, fishmonger, butchery, charcuterie, dairy and waste collection industries.
Specifications
Storage Tote | |
White | |
15.9 gal. |
HDPE | |
Rectangular | |
60 L |
Product Content Correction
Product Title
Gilac™ Reinforced Stacking and Nesting Container > Reinforced Stacking and Nesting Container; Volume: 60 L; Color: White
Spot an opportunity for improvement?Share a Content Correction