missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Reinforced Stacking and Nesting Container

Reinforced Stacking and Nesting Container is designed for intensive use and extreme handling conditions; it's stackable and nestable by a quarter turn.

902.00 DKK - 1120.00 DKK

Specifications

Type Storage Tote
Material HDPE
Color White
Shape Rectangular
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
17998861
View Documents
Gilac™
G169321
60 L
902.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
 
17938671
View Documents
Gilac™
G170521
100 L
1120.00 DKK
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
 
Description

Description

  • Reinforced containers for intensive use and extreme handling conditions.
  • Stackable and nestable by a quarter turn.
  • Highly durable.
  • Suitable for a number of different uses in the food processing, fishmonger, butchery, charcuterie, dairy and waste collection industries.
Specifications

Specifications

Storage Tote
HDPE
White
Rectangular
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilac™ Reinforced Stacking and Nesting Container

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.