Learn More
Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths
Designed specifically to generate maximum ultrasonic activity. Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths prevent operators coming into contact with chemical solutions.
Brand: Grant Instruments™ XAB3
880.39 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Stainless steel construction
- Works in conjunction with lid to create a drip collection unit
- Prevents user from coming into contact with chemical solutions
- Prevents items from resting on the inside of the tank wall
- For use with Grant Ultrasonic Baths
Specifications
XUBA3 Ultrasonic Bath | |
Stainless Steel | |
(WxDxH) 208 x 115 x 98 mm |