Learn More
Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths
Designed specifically to generate maximum ultrasonic activity. Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths prevent operators coming into contact with chemical solutions.
697.00 DKK - 2295.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
11401018
|
Grant Instruments™
XAB12 |
XUB12 Ultrasonic Bath |
1595.00 DKK
Each |
|
||||||
11411018
|
Grant Instruments™
XAB18 |
XUB18 Ultrasonic Bath |
2070.00 DKK
Each |
|
||||||
11421018
|
Grant Instruments™
XAB25 |
XUB25 Ultrasonic Bath |
2295.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
11481008
|
Grant Instruments™
XAB5 |
XUB5 Ultrasonic Bath |
1010.00 DKK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
11461008
|
Grant Instruments™
XAB1 |
XUBA1 Ultrasonic Bath |
697.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11471008
|
Grant Instruments™
XAB3 |
XUBA3 Ultrasonic Bath |
908.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
Description
- Stainless steel construction
- Works in conjunction with lid to create a drip collection unit
- Prevents user from coming into contact with chemical solutions
- Prevents items from resting on the inside of the tank wall
- For use with Grant Ultrasonic Baths
Specifications
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths