Learn More
Benchmark Scientific™ RotoBot™ Rotating Mixers
RotoBot™ Rotating Mixers feature advanced programming options, digital speed control and more than 10 operating modes. A wide variety of rotating, mixing and vibrating actions can be combined to provide the precise agitation profiles. Supplied with holders for all common tubes from 1.5 to 50 mL.
Brand:  Benchmark Scientific™ R4045-E
 View more versions of this product
Product Code. 17994432
  • 8290.00 DKK / Each
Explore more special offers
Specifications
360°
3 A
1.5 to 50 mL
Digital
100/230 V
Tubes from 1.5 mL to 50 mL
UK
6 x 1.5 mL Holder (5), 1 x 15 mL Holder (8), 1 x 50 mL Holder (2)