missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPLP0 Mouse anti-Human, Mouse, Rat, Clone: 2A2E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68056-1-IG-150UL
This item is not returnable.
View return policy
Description
RPLP0 Monoclonal antibody apecifically detetects RPLP0 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
RPLP0 | |
Monoclonal | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
RPLP0 | |
RPLP0 fusion protein Ag1829 | |
150 μL | |
6175 | |
Store at -20°C. Stable for one year after shipment. | |
IgG1 |
Western Blot, Immunohistochemistry, ELISA | |
2A2E1 | |
Unconjugated | |
BC009867 | |
Mouse | |
Protein G purification | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPLP0 Mouse anti-Human, Mouse, Rat, Clone: 2A2E1, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction