missing translation for 'onlineSavingsMsg'
Learn More

RPLP0 Mouse anti-Human, Mouse, Rat, Clone: 2A2E1, Proteintech

Mouse Monoclonal Antibody

1081.84 DKK - 3058.99 DKK

Specifications

Antigen RPLP0
Clone 2A2E1
Concentration 1000 ÎĽg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17669716
View Documents
Proteintech
68056-1-IG-20UL
20 ÎĽL
1081.84 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
17659716
View Documents
Proteintech
68056-1-IG-150UL
150 ÎĽL
3058.99 DKK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RPLP0 Monoclonal antibody apecifically detetects RPLP0 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.
Specifications

Specifications

RPLP0
1000 ÎĽg/mL
Monoclonal
Liquid
Human, Mouse, Rat
BC009867
RPLP0
IgG1
Protein G purification
2A2E1
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Mouse
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
6175
RPLP0 fusion protein Ag1829
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RPLP0 Mouse anti-Human, Mouse, Rat, Clone: 2A2E1, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.