missing translation for 'onlineSavingsMsg'
Learn More

RPLP0 Mouse anti-Human, Mouse, Rat, Clone: 2A2E1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68056-1-IG-20UL

 View more versions of this product

Product Code. 17669716

  • 1081.84 DKK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RPLP0 Monoclonal antibody apecifically detetects RPLP0 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.
Specifications

Specifications

RPLP0
Monoclonal
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
RPLP0
RPLP0 fusion protein Ag1829
20 μL
6175
Store at -20°C. Stable for one year after shipment.
IgG1
Western Blot, Immunohistochemistry, ELISA
2A2E1
Unconjugated
BC009867
Mouse
Protein G purification
Primary
Human, Mouse, Rat
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RPLP0 Mouse anti-Human, Mouse, Rat, Clone: 2A2E1, Proteintech > 20 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.