Learn More
Fisherbrand™ Safety Cover for Dry Block Heaters
For use with Fisherbrand™ Dry Block Heaters.
Brand:  Fisherbrand™ 17571896
 View more versions of this product
Product Code. 17571896
  • 1055.00 DKK / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Description
  • Made from tough clear acrylic for maximum visibility
  • Prevents accidental touching of a hot block or contamination of samples from splashes
Specifications
Specifications
Fisherbrand™ 2 Block System
Preventing Burns or Contamination
85 mm
Safety Cover
Acrylic